Assay Details
Target Gene Details
Entrez Gene ID: | 79611 |
Gene Name: | acyl-CoA synthetase short-chain family member 3 |
Gene Aliases: |
- |
Location: |
Chr.12:81078018-81257795 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACSS3 | NM_024560.2 | NP_078836.1 | ||
| XM_005269150.2 | XP_005269207.1 | |||
| XM_005269151.4 | XP_005269208.1 | |||
| AK025616.1 | BAB15190.1 | |||
| AX764480.1 | CAE00104.1 | |||
| AX764482.1 | CAE00105.1 | |||
| BC009317.2 | AAH09317.1 | |||
| BC015769.1 | AAH15769.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892231 | Chr.12:81076082 - 81120636 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map