Assay Details
Target Gene Details
Entrez Gene ID: | 84766 |
Gene Name: | calcium release activated channel regulator 2A |
Gene Aliases: |
EFCAB4B |
Location: |
Chr.12:3613366-3753216 on Build GRCh38 |
Assay Gene Location: | Within Intron 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRACR2A | NM_001144958.1 | NP_001138430.1 | ||
| XM_006719021.3 | XP_006719084.1 | |||
| XM_011521034.2 | XP_011519336.1 | |||
| XM_011521036.2 | XP_011519338.1 | |||
| XM_011521037.2 | XP_011519339.1 | |||
| XM_011521038.2 | XP_011519340.1 | |||
| AK091124.1 | ||||
| AK304017.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv557116 | Chr.12:3576597 - 3989342 on Build GRCh38 | Gain |
|
| nsv582 | Chr.12:3596830 - 3627639 on Build GRCh38 | Insertion |
|
| nsv483060 | Chr.12:3190835 - 5290834 on Build GRCh38 | Loss |
|
| nsv1036828 | Chr.12:3607219 - 3846326 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map