Assay Details
Target Gene Details
Entrez Gene ID: | 10445 |
Gene Name: | microspherule protein 1 |
Gene Aliases: |
ICP22BP, INO80Q, MCRS2, MSP58, P78 |
Location: |
Chr.12:49558294-49572157 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MCRS1 | XM_011537760.2 | 1 | 3081 | XP_011536062.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832406 | Chr.12:49466910 - 49668353 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map