Assay Details
Target Gene Details
Entrez Gene ID: | 144608 |
Gene Name: | chromosome 12 open reading frame 60 |
Gene Aliases: |
- |
Location: |
Chr.12:14567991-14823858 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C12orf60 | XM_017018871.1 | XP_016874360.1 | ||
| XM_017018872.1 | XP_016874361.1 |
Target Gene Details
Entrez Gene ID: | 51729 |
Gene Name: | WW domain binding protein 11 |
Gene Aliases: |
NPWBP, PPP1R165, SIPP1, WBP-11 |
Location: |
Chr.12:14786478-14803467 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WBP11 | NM_016312.2 | NP_057396.1 | ||
| AB029309.1 | BAA88410.1 | |||
| AF118023.1 | AAD30425.1 | |||
| AK297410.1 | ||||
| AK302244.1 | ||||
| BC001621.1 | AAH01621.1 | |||
| BC016441.2 | AAH16441.2 | |||
| BC023532.2 | AAH23532.1 | |||
| BX647347.1 |
More Information
Set Membership: |
|

Genomic Map