Assay Details
Target Gene Details
Entrez Gene ID: | 775 |
Gene Name: | calcium voltage-gated channel subunit alpha1 C |
Gene Aliases: |
CACH2, CACN2, CACNL1A1, CCHL1A1, CaV1.2, LQT8, TS |
Location: |
Chr.12:1969677-2697949 on Build GRCh38 |
Assay Gene Location: | Within Intron 46 |
Target Gene Details
Entrez Gene ID: | 100652846 |
Gene Name: | CACNA1C antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.12:2676001-2691157 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CACNA1C-AS1 | NR_045725.1 | 4 | 1761 | |
| AK093746.1 | 4 | 1722 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1047874 | Chr.12:2577599 - 2807322 on Build GRCh38 | Gain |
|
| nsv832308 | Chr.12:2576562 - 2690062 on Build GRCh38 | Loss |
|
| dgv1344n100 | Chr.12:2673632 - 2740600 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map