Assay Details
Target Gene Details
Entrez Gene ID: | 105369914 |
Gene Name: | translation initiation factor IF-2-like |
Gene Aliases: |
- |
Location: |
Chr.12:94648482-94651098 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105369914 | XM_011539068.2 | 1 | 498 | XP_011537370.1 |
Target Gene Details
Entrez Gene ID: | 57458 |
Gene Name: | transmembrane and coiled-coil domain family 3 |
Gene Aliases: |
- |
Location: |
Chr.12:94567124-94650562 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMCC3 | NM_020698.3 | NP_065749.2 | ||
| BC040535.1 | AAH40535.1 |
More Information
Set Membership: |
|

Genomic Map