Assay Details
Target Gene Details
Entrez Gene ID: | 90627 |
Gene Name: | StAR related lipid transfer domain containing 13 |
Gene Aliases: |
ARHGAP37, DLC2, GT650, LINC00464 |
Location: |
Chr.13:33103135-33676835 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STARD13 | NM_001243476.2 | NP_001230405.1 | ||
| NM_178006.3 | NP_821074.1 | |||
| XM_011535299.2 | XP_011533601.1 | |||
| XM_017020834.1 | XP_016876323.1 | |||
| XM_017020835.1 | XP_016876324.1 | |||
| AK094704.1 | ||||
| AK308430.1 | ||||
| AK308453.1 | ||||
| AK312888.1 | ||||
| AY082589.1 | AAL91648.1 | |||
| AY082592.1 | AAL91651.1 | |||
| AY366448.1 | AAQ72791.1 | |||
| BX647695.1 | CAI46026.1 | |||
| DA312208.1 | ||||
| DC402824.1 |
Target Gene Details
Entrez Gene ID: | 100874241 |
Gene Name: | STARD13 antisense RNA |
Gene Aliases: |
STARD13-AS2 |
Location: |
Chr.13:33277554-33281334 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STARD13-AS | NR_046693.1 | 3 | 438 | |
| AL137709.1 | 1 | 1198 | ||
| BX103803.1 | 3 | 438 |
More Information
Set Membership: |
|

Genomic Map