Assay Details
Target Gene Details
Entrez Gene ID: | 79866 |
Gene Name: | bora, aurora kinase A activator |
Gene Aliases: |
C13orf34 |
Location: |
Chr.13:72727739-72756198 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BORA | NM_001286746.1 | NP_001273675.1 | ||
| NM_001286747.1 | NP_001273676.1 | |||
| NM_024808.3 | NP_079084.3 | |||
| XM_006719868.3 | XP_006719931.2 | |||
| AK291496.1 | ||||
| AK298608.1 | ||||
| AK300758.1 | ||||
| BC025367.2 | AAH25367.2 | |||
| BC056876.1 | AAH56876.2 | |||
| BM923278.1 | ||||
| BX648605.1 | ||||
| EU834129.1 |
More Information
Set Membership: |
|

Genomic Map