Assay Details
Target Gene Details
Entrez Gene ID: | 221143 |
Gene Name: | eukaryotic translation elongation factor 1 alpha lysine methyltransferase 1 |
Gene Aliases: |
ESP13, N6AMT2 |
Location: |
Chr.13:20728931-20773958 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EEF1AKMT1 | NM_001318939.1 | NP_001305868.1 | ||
| NM_174928.2 | NP_777588.1 | |||
| NR_134934.1 | ||||
| AK316604.1 | ||||
| AL597222.1 | ||||
| BC018091.1 | AAH18091.1 | |||
| DA514874.1 | ||||
| DQ823637.1 | ||||
| HY004043.1 | 1 | 397 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv951 | Chr.13:20753248 - 20779879 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map