Assay Details
Target Gene Details
Entrez Gene ID: | 160897 |
Gene Name: | G protein-coupled receptor 180 |
Gene Aliases: |
ITR |
Location: |
Chr.13:94601850-94634645 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR180 | NM_180989.5 | NP_851320.1 | ||
| AK126794.1 | BAC86697.1 | |||
| AK289850.1 | ||||
| AL832739.1 | 1 | 1353 | CAI46124.1 | |
| BC052243.1 | AAH52243.1 | |||
| BI856811.1 | ||||
| DB150630.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832686 | Chr.13:94554488 - 94748738 on Build GRCh38 | Gain |
|
| nsv832685 | Chr.13:94527422 - 94627712 on Build GRCh38 | Gain |
|
| esv34004 | Chr.13:94254283 - 94740390 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map