Assay Details
Target Gene Details
Entrez Gene ID: | 5101 |
Gene Name: | protocadherin 9 |
Gene Aliases: |
- |
Location: |
Chr.13:66302834-67230336 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDH9 | NM_001318372.1 | NP_001305301.1 | ||
| NM_001318373.1 | NP_001305302.1 | |||
| NM_020403.4 | NP_065136.1 | |||
| NM_203487.2 | NP_982354.1 | |||
| XM_017020619.1 | XP_016876108.1 | |||
| XM_017020620.1 | XP_016876109.1 | |||
| AF169692.2 | AAF89689.2 | |||
| AL834258.1 | CAD38933.1 | |||
| BC136626.1 | ||||
| BC136627.1 | ||||
| BC144321.1 | ||||
| BX537422.1 | CAD97664.1 | |||
| KJ534915.1 |
Target Gene Details
Entrez Gene ID: | 100874086 |
Gene Name: | PCDH9 antisense RNA 3 |
Gene Aliases: |
- |
Location: |
Chr.13:66977389-66985776 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDH9-AS3 | NR_046636.1 | 3 | 436 | |
| AI073488.1 | 1 | 90 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832640 | Chr.13:66886172 - 67061834 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map