Assay Details
Target Gene Details
Entrez Gene ID: | 101928922 |
Gene Name: | long intergenic non-protein coding RNA 463 |
Gene Aliases: |
AMER2-AS1 |
Location: |
Chr.13:25172824-25180001 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC00463 | NR_120428.1 | |||
| BU182247.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3582237 | Chr.13:25162386 - 25434284 on Build GRCh38 | Gain |
|
| esv2747161 | Chr.13:25031743 - 26209531 on Build GRCh38 | Deletion |
|

Genomic Map