Assay Details
Target Gene Details
Entrez Gene ID: | 10443 |
Gene Name: | NEDD4 binding protein 2 like 2 |
Gene Aliases: |
92M18.3, CG005, CG016, PFAAP5 |
Location: |
Chr.13:32432488-32542710 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Intron 13 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832576 | Chr.13:32322994 - 32502851 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map