Assay Details
Target Gene Details
Entrez Gene ID: | 100507398 |
Gene Name: | INTS6 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.13:51453345-51454716 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INTS6-AS1 | NR_103812.1 | 2 | 478 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892343 | Chr.13:51418581 - 52060832 on Build GRCh38 | Gain |
|
| nsv561618 | Chr.13:49907577 - 51521449 on Build GRCh38 | Loss |
|

Genomic Map