Assay Details
Target Gene Details
Entrez Gene ID: | 267012 |
Gene Name: | D-amino acid oxidase activator |
Gene Aliases: |
LG72, SG72 |
Location: |
Chr.13:105465867-105491034 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DAOA | NM_001161812.1 | NP_001155284.1 | ||
| NM_001161814.1 | NP_001155286.1 | |||
| NM_172370.4 | NP_758958.3 | |||
| XM_005254042.1 | XP_005254099.1 | |||
| AY138546.1 | AAN08432.1 | |||
| AY138547.1 | AAN08433.1 | |||
| AY170469.2 | AAO12727.1 | |||
| AY170470.2 | AAO12728.1 | |||
| AY170471.1 | AAO12729.1 | |||
| AY223901.1 | AAO73604.1 | |||
| BC121091.1 | ||||
| DQ343761.1 | ||||
| DQ357223.1 | ||||
| DQ386869.1 | ||||
| DQ386870.1 |
Target Gene Details
Entrez Gene ID: | 282706 |
Gene Name: | DAOA antisense RNA 1 |
Gene Aliases: |
DAOA-AS, DAOAAS, G30 |
Location: |
Chr.13:105459055-105505681 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DAOA-AS1 | NR_040247.1 | |||
| AY138548.1 | AAN08434.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv562996 | Chr.13:104426430 - 105664629 on Build GRCh38 | Gain |
|
| nsv519018 | Chr.13:105467097 - 105473959 on Build GRCh38 | Loss |
|
| nsv1048036 | Chr.13:105463559 - 105534128 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map