Assay Details
Target Gene Details
Entrez Gene ID: | 10464 |
Gene Name: | progesterone immunomodulatory binding factor 1 |
Gene Aliases: |
C13orf24, CEP90, PIBF |
Location: |
Chr.13:72782059-73016454 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIBF1 | NM_006346.2 | NP_006337.2 | ||
| XM_011534881.2 | XP_011533183.1 | |||
| XM_011534882.2 | XP_011533184.1 | |||
| XM_011534884.2 | XP_011533186.1 | |||
| XM_011534885.2 | XP_011533187.1 | |||
| XM_011534886.2 | XP_011533188.1 | |||
| XM_017020350.1 | XP_016875839.1 | |||
| XM_017020351.1 | XP_016875840.1 | |||
| XM_017020352.1 | XP_016875841.1 | |||
| XM_017020353.1 | XP_016875842.1 | |||
| XM_017020354.1 | XP_016875843.1 | |||
| AF330046.1 | AAL37481.1 | |||
| AK309828.1 | ||||
| AY370776.1 | AAQ73282.1 | |||
| AY375528.1 | AAQ67659.1 | |||
| BC006009.1 | ||||
| BC041806.1 | AAH41806.1 | |||
| BC051911.1 | AAH51911.1 | |||
| BX341357.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1049859 | Chr.13:72741128 - 72872273 on Build GRCh38 | Loss |
|
| esv3580764 | Chr.13:72767884 - 72798860 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map