Assay Details
Target Gene Details
Entrez Gene ID: | 51082 |
Gene Name: | RNA polymerase I subunit D |
Gene Aliases: |
AC19, POLR1C, RPA16, RPA9, RPAC2, RPC16, RPO1-3, TCS2 |
Location: |
Chr.13:27620743-27667422 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLR1D | NM_001206559.1 | NP_001193488.1 | ||
| NM_152705.2 | NP_689918.1 | |||
| XM_005266414.2 | XP_005266471.1 | |||
| XM_017020623.1 | XP_016876112.1 | |||
| AK097973.1 | ||||
| AK307356.1 | ||||
| AK311833.1 | ||||
| BC006972.1 | AAH06972.1 | |||
| BC015319.2 | AAH15319.1 | |||
| BC018528.1 | AAH18528.1 | |||
| DB123903.1 | ||||
| DB455307.1 | ||||
| DB459107.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1039183 | Chr.13:27600204 - 27632386 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map