Assay Details
Target Gene Details
Entrez Gene ID: | 3651 |
Gene Name: | pancreatic and duodenal homeobox 1 |
Gene Aliases: |
GSF, IDX-1, IPF1, IUF1, MODY4, PAGEN1, PDX-1, STF-1 |
Location: |
Chr.13:27919982-27926314 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDX1 | NM_000209.3 | 2 | 2206 | NP_000200.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv538e212 | Chr.13:27922749 - 27955049 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map