Assay Details
Target Gene Details
Entrez Gene ID: | 219402 |
Gene Name: | mitochondrial translational initiation factor 3 |
Gene Aliases: |
IF3mt |
Location: |
Chr.13:27435639-27450900 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTIF3 | XM_006719772.3 | 1 | 148 | XP_006719835.1 |
| XM_011534957.2 | 1 | 146 | XP_011533259.1 | |
| XM_011534961.2 | 1 | 146 | XP_011533263.1 | |
| XM_011534963.2 | 1 | 146 | XP_011533265.1 |
More Information
Set Membership: |
|

Genomic Map