Assay Details
Target Gene Details
Entrez Gene ID: | 100874161 |
Gene Name: | long intergenic non-protein coding RNA 411 |
Gene Aliases: |
- |
Location: |
Chr.13:100939520-100944364 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC00411 | NR_047015.1 | |||
| AA846322.1 | ||||
| AI183969.1 |
Target Gene Details
Entrez Gene ID: | 100885778 |
Gene Name: | NALCN antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.13:100708325-101059286 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NALCN-AS1 | NR_047687.1 | |||
| AK025806.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv562958 | Chr.13:100910655 - 100952196 on Build GRCh38 | Loss |
|

Genomic Map