Assay Details
Target Gene Details
Entrez Gene ID: | 253512 |
Gene Name: | solute carrier family 25 member 30 |
Gene Aliases: |
KMCP1 |
Location: |
Chr.13:45393316-45434016 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC25A30 | XM_017020523.1 | XP_016876012.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv223e201 | Chr.13:45377870 - 46464067 on Build GRCh38 | Deletion |
|
| nsv510579 | Chr.13:45305226 - 45424938 on Build GRCh38 | Deletion |
|
| esv3631957 | Chr.13:45413790 - 45431769 on Build GRCh38 | Loss |
|
| nsv561578 | Chr.13:45415321 - 45479409 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map