Assay Details
Target Gene Details
Entrez Gene ID: | 219333 |
Gene Name: | ubiquitin specific peptidase 12 |
Gene Aliases: |
UBH1, USP12L1 |
Location: |
Chr.13:27066150-27171896 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP12 | NM_182488.3 | NP_872294.2 | ||
| XM_005266282.2 | XP_005266339.1 | |||
| XM_011534956.2 | XP_011533258.1 | |||
| XM_017020416.1 | XP_016875905.1 | |||
| AF022789.1 | AAC23551.1 | |||
| AK289685.1 | ||||
| BC026072.1 | AAH26072.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs74041148] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map