Assay Details
Target Gene Details
Entrez Gene ID: | 22894 |
Gene Name: | DIS3 homolog, exosome endoribonuclease and 3'-5' exoribonuclease |
Gene Aliases: |
2810028N01Rik, EXOSC11, KIAA1008, RRP44, dis3p |
Location: |
Chr.13:72755402-72782128 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DIS3 | NM_001128226.2 | NP_001121698.1 | ||
| NM_001322348.1 | NP_001309277.1 | |||
| NM_001322349.1 | NP_001309278.1 | |||
| NM_014953.4 | NP_055768.3 | |||
| AB001743.1 | ||||
| AB023225.1 | ||||
| AF330044.1 | AAL37479.1 | |||
| AK025634.1 | ||||
| AK314715.1 | ||||
| AL080158.1 | CAB45749.1 | |||
| AL832266.1 | CAH56266.1 | |||
| BC038101.1 | AAH38101.1 | |||
| BC056143.1 | AAH56143.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1049859 | Chr.13:72741128 - 72872273 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map