Assay Details
Target Gene Details
Entrez Gene ID: | 81033 |
Gene Name: | potassium voltage-gated channel subfamily H member 6 |
Gene Aliases: |
ERG-2, ERG2, HERG2, Kv11.2, hERG-2 |
Location: |
Chr.17:63523334-63548977 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3640998 | Chr.17:63526371 - 63539713 on Build GRCh38 | Loss |
|
| nsv952355 | Chr.17:63522640 - 63550539 on Build GRCh38 | Deletion |
|
| esv2675657 | Chr.17:63526487 - 63539627 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map