Assay Details
Target Gene Details
Entrez Gene ID: | 57602 |
Gene Name: | ubiquitin specific peptidase 36 |
Gene Aliases: |
DUB1 |
Location: |
Chr.17:78787381-78840898 on Build GRCh38 |
Assay Gene Location: | Within Intron 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP36 | XM_011525067.2 | XP_011523369.1 | ||
| XM_011525071.2 | XP_011523373.1 | |||
| XM_017024900.1 | XP_016880389.1 | |||
| AB040886.1 | ||||
| AK022913.1 | BAB14306.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520359 | Chr.17:78782658 - 78788699 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map