Assay Details
Target Gene Details
Entrez Gene ID: | 10140 |
Gene Name: | transducer of ERBB2, 1 |
Gene Aliases: |
APRO5, APRO6, PIG49, TOB, TROB, TROB1 |
Location: |
Chr.17:50862223-50867978 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TOB1 | NM_001243877.1 | NP_001230806.1 | ||
| NM_001243885.1 | NP_001230814.1 | |||
| NM_005749.3 | NP_005740.1 | |||
| AA675891.1 | ||||
| AK313904.1 | ||||
| BC070493.1 | AAH70493.1 | |||
| BC098415.1 | AAH98415.1 | |||
| DA039934.1 |
More Information
Set Membership: |
|

Genomic Map