Assay Details
Target Gene Details
Entrez Gene ID: | 8787 |
Gene Name: | regulator of G-protein signaling 9 |
Gene Aliases: |
PERRS, RGS9L |
Location: |
Chr.17:65137338-65227703 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RGS9 | NM_001081955.2 | NP_001075424.1 | ||
| NM_001165933.1 | NP_001159405.1 | |||
| NM_003835.3 | NP_003826.2 | |||
| AF071476.1 | AAC64040.1 | |||
| AF493933.1 | AAM12647.1 | |||
| AK289876.1 | ||||
| AK290535.1 | ||||
| AK301719.1 | ||||
| AK309061.1 | ||||
| AY585191.1 | AAT79494.1 | |||
| BC022504.1 | AAH22504.1 | |||
| CD105605.1 | ||||
| DC322215.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv1009898 | Chr.17:65139616 - 65150605 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map