Assay Details
Target Gene Details
Entrez Gene ID: | 101928514 |
Gene Name: | uncharacterized LOC101928514 |
Gene Aliases: |
- |
Location: |
Chr.17:76799044-76807102 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928514 | NR_110837.1 | |||
| AK055500.1 |
Target Gene Details
Entrez Gene ID: | 79157 |
Gene Name: | major facilitator superfamily domain containing 11 |
Gene Aliases: |
ET |
Location: |
Chr.17:76736565-76803805 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MFSD11 | XM_011525242.1 | XP_011523544.1 | ||
| XM_011525244.1 | XP_011523546.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833550 | Chr.17:76712038 - 76886290 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map