Assay Details
Target Gene Details
Entrez Gene ID: | 7266 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member C7 |
Gene Aliases: |
DJ11, DJC7, TPR2, TTC2 |
Location: |
Chr.17:41976421-42021371 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJC7 | XM_011525168.2 | XP_011523470.1 |
Target Gene Details
Entrez Gene ID: | 28511 |
Gene Name: | NFKB inhibitor interacting Ras like 2 |
Gene Aliases: |
KBRAS2, kappaB-Ras2 |
Location: |
Chr.17:42017576-42025641 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NKIRAS2 | NM_001001349.2 | NP_001001349.1 | ||
| NM_001144927.1 | NP_001138399.1 | |||
| NM_001144928.1 | NP_001138400.1 | |||
| NM_001144929.1 | NP_001138401.1 | |||
| NM_017595.5 | NP_060065.2 | |||
| XM_005257251.1 | XP_005257308.1 | |||
| AK027265.1 | BAB55006.1 | |||
| AK054571.1 | ||||
| AK297975.1 | ||||
| AL137682.1 | 1 | 173 | CAB70873.2 | |
| BC007450.2 | AAH07450.1 | |||
| BC063498.1 | AAH63498.1 | |||
| BG717103.1 | ||||
| DA509824.1 | ||||
| DA607452.1 |
More Information
Set Membership: |
|

Genomic Map