Assay Details
Target Gene Details
Entrez Gene ID: | 57551 |
Gene Name: | TAO kinase 1 |
Gene Aliases: |
KFC-B, MAP3K16, MARKK, PSK-2, PSK2, TAO1, hKFC-B, hTAOK1 |
Location: |
Chr.17:29390925-29551904 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAOK1 | NM_020791.2 | NP_065842.1 | ||
| NM_025142.1 | NP_079418.1 | |||
| BC133039.1 | ||||
| BC144067.1 | ||||
| BE772351.1 | ||||
| CA395289.1 |
More Information
Set Membership: |
|

Genomic Map