Assay Details
Target Gene Details
Entrez Gene ID: | 10140 |
Gene Name: | transducer of ERBB2, 1 |
Gene Aliases: |
APRO5, APRO6, PIG49, TOB, TROB, TROB1 |
Location: |
Chr.17:50862223-50867978 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TOB1 | NM_001243877.1 | NP_001230806.1 | ||
| AK313904.1 |
Target Gene Details
Entrez Gene ID: | 400604 |
Gene Name: | TOB1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.17:50866679-50868371 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TOB1-AS1 | NR_038458.1 | |||
| BP421960.1 | ||||
| BQ222352.1 |
More Information
Set Membership: |
|

Genomic Map