Assay Details
Target Gene Details
Entrez Gene ID: | 6198 |
Gene Name: | ribosomal protein S6 kinase B1 |
Gene Aliases: |
PS6K, S6K, S6K-beta-1, S6K1, STK14A, p70 S6KA, p70(S6K)-alpha, p70-S6K, p70-alpha |
Location: |
Chr.17:59893046-59950426 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPS6KB1 | NM_001272042.1 | NP_001258971.1 | ||
| NM_001272043.1 | NP_001258972.1 | |||
| NM_001272044.1 | NP_001258973.1 | |||
| NM_001272060.1 | NP_001258989.1 | |||
| NM_003161.3 | NP_003152.1 | |||
| XM_011525101.2 | XP_011523403.1 | |||
| XM_011525102.2 | XP_011523404.1 | |||
| XM_011525103.2 | XP_011523405.1 | |||
| XM_017024929.1 | XP_016880418.1 | |||
| XM_017024930.1 | XP_016880419.1 | |||
| XM_017024931.1 | XP_016880420.1 | |||
| XM_017024932.1 | XP_016880421.1 | |||
| XM_017024933.1 | XP_016880422.1 | |||
| AK293247.1 | ||||
| AK297147.1 | ||||
| AK300202.1 | ||||
| AK312875.1 | ||||
| BC036033.1 | AAH36033.1 | |||
| BC053365.1 | AAH53365.1 | |||
| BP359158.1 | ||||
| M60724.1 | AAA36410.1 |
More Information
Set Membership: |
|

Genomic Map