Assay Details
Target Gene Details
Entrez Gene ID: | 5709 |
Gene Name: | proteasome 26S subunit, non-ATPase 3 |
Gene Aliases: |
P58, RPN3, S3, TSTA2 |
Location: |
Chr.17:39980768-39997960 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PSMD3 | NM_002809.3 | NP_002800.2 | ||
| AK022896.1 | ||||
| AK054611.1 | ||||
| AK055799.1 | BAB71019.1 | |||
| AK094206.1 | BAC04310.1 | |||
| AK293539.1 | ||||
| AK298411.1 | ||||
| AK300081.1 | ||||
| AK300973.1 | ||||
| BC000074.2 | AAH00074.1 | |||
| BC004859.2 | AAH04859.1 | |||
| BC012302.1 | AAH12302.1 | |||
| BC020518.1 | AAH20518.1 | |||
| BC025686.1 | AAH25686.1 | |||
| BT007217.1 | AAP35881.1 | |||
| CR456772.1 | CAG33053.1 | |||
| D67025.1 | BAA23651.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3573064 | Chr.17:39896348 - 40029806 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map