Assay Details
Target Gene Details
Entrez Gene ID: | 9097 |
Gene Name: | ubiquitin specific peptidase 14 |
Gene Aliases: |
TGT |
Location: |
Chr.18:158483-213739 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP14 | NM_001037334.1 | NP_001032411.1 | ||
| NM_005151.3 | NP_005142.1 | |||
| AK095516.1 | ||||
| AK297605.1 | ||||
| AK315438.1 | ||||
| BC003556.1 | AAH03556.1 | |||
| BG717018.1 | ||||
| BT007183.1 | AAP35847.1 | |||
| CR976282.1 | ||||
| U30888.1 | AAB60365.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv576227 | Chr.18:52933 - 322183 on Build GRCh38 | Gain |
|
| esv3641475 | Chr.18:153985 - 282924 on Build GRCh38 | Gain |
|
| dgv3301n100 | Chr.18:11543 - 326679 on Build GRCh38 | Gain |
|
| nsv1064150 | Chr.18:99036 - 321710 on Build GRCh38 | Gain |
|
| nsv482935 | Chr.18:10001 - 2900002 on Build GRCh38 | Loss |
|
| nsv576228 | Chr.18:69836 - 1506323 on Build GRCh38 | Loss |
|
| esv3641472 | Chr.18:128077 - 323706 on Build GRCh38 | Loss |
|
| dgv3302n100 | Chr.18:136228 - 249310 on Build GRCh38 | Gain |
|
| esv3573886 | Chr.18:136227 - 218394 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115281583] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map