Assay Details
Target Gene Details
Entrez Gene ID: | 652991 |
Gene Name: | SKI family transcriptional corepressor 2 |
Gene Aliases: |
CH18515, CORL2, FUSSEL18 |
Location: |
Chr.18:47206182-47249524 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SKOR2 | NM_001278063.1 | NP_001264992.1 | ||
| XM_006722525.3 | XP_006722588.1 | |||
| DW009659.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9629 | Chr.18:46684730 - 47231140 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map