Assay Details
Target Gene Details
Entrez Gene ID: | 100505592 |
Gene Name: | gastric adenocarcinoma associated, positive CD44 regulator, long intergenic non-coding RNA |
Gene Aliases: |
LINC01540 |
Location: |
Chr.18:3466250-3478978 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GAPLINC | NR_110428.1 | |||
| NR_110429.1 | ||||
| AJ707365.1 | ||||
| BM782281.1 | ||||
| BX107736.1 |

Genomic Map