Assay Details
Target Gene Details
Entrez Gene ID: | 10982 |
Gene Name: | microtubule associated protein RP/EB family member 2 |
Gene Aliases: |
CSCSC2, EB1, EB2, RP1 |
Location: |
Chr.18:34976928-35143470 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAPRE2 | NM_001143826.2 | NP_001137298.1 | ||
| NM_001143827.2 | NP_001137299.1 | |||
| AK294833.1 | ||||
| DC335558.1 | ||||
| DC341673.1 |
More Information
Set Membership: |
|

Genomic Map