Assay Details
Target Gene Details
Entrez Gene ID: | 10449 |
Gene Name: | acetyl-CoA acyltransferase 2 |
Gene Aliases: |
DSAEC |
Location: |
Chr.18:49783504-49813881 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACAA2 | NM_006111.2 | NP_006102.2 | ||
| AK054673.1 | ||||
| AK225457.1 | ||||
| AK314462.1 | ||||
| BC001918.1 | AAH01918.1 | |||
| D16294.1 | BAA03800.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv516056 | Chr.18:49768512 - 49838635 on Build GRCh38 | Gain |
|
| nsv576863 | Chr.18:49569117 - 49938639 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map