Assay Details
Target Gene Details
Entrez Gene ID: | 5271 |
Gene Name: | serpin family B member 8 |
Gene Aliases: |
CAP2, PI8 |
Location: |
Chr.18:63970028-64019779 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SERPINB8 | NM_001031848.1 | NP_001027018.1 | ||
| NM_001276490.1 | NP_001263419.1 | |||
| NM_002640.3 | NP_002631.3 | |||
| NM_198833.1 | NP_942130.1 | |||
| XM_006722484.2 | XP_006722547.1 | |||
| XM_011526025.1 | XP_011524327.1 | |||
| XM_017025790.1 | XP_016881279.1 | |||
| XM_017025791.1 | XP_016881280.1 | |||
| XM_017025792.1 | XP_016881281.1 | |||
| AK300391.1 | ||||
| AK313782.1 | ||||
| AL708543.1 | ||||
| BC034528.1 | AAH34528.1 | |||
| BQ435307.1 | ||||
| BX280409.1 | ||||
| BX571754.1 | CAE11879.1 | |||
| DC384755.1 | ||||
| DQ656055.2 | ||||
| L40377.1 | AAC41939.1 | |||
| U92984.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1066552 | Chr.18:63147299 - 64608176 on Build GRCh38 | Gain |
|
| nsv833673 | Chr.18:63897072 - 64097021 on Build GRCh38 | Gain |
|
| esv21739 | Chr.18:63715122 - 64278129 on Build GRCh38 | Gain |
|
| esv3642785 | Chr.18:63831255 - 63986077 on Build GRCh38 | Gain |
|
| nsv1067311 | Chr.18:63818011 - 66637685 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map