Assay Details
Target Gene Details
Entrez Gene ID: | 64863 |
Gene Name: | methyltransferase like 4 |
Gene Aliases: |
HsT661 |
Location: |
Chr.18:2537525-2571503 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| METTL4 | NM_001308401.1 | NP_001295330.1 | ||
| NM_022840.4 | NP_073751.3 | |||
| XM_005258132.3 | XP_005258189.1 | |||
| XM_005258133.2 | XP_005258190.1 | |||
| XM_011525730.2 | XP_011524032.1 | |||
| AK022584.1 | ||||
| AK026670.1 | BAB15520.1 | |||
| AK290001.1 | ||||
| AL834296.1 | CAD38969.1 | |||
| BC111020.1 | AAI11021.1 | |||
| BC136766.1 | ||||
| BC136767.1 | ||||
| BX647373.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1061548 | Chr.18:2163157 - 2631423 on Build GRCh38 | Gain |
|
| nsv1065830 | Chr.18:2263727 - 2576042 on Build GRCh38 | Gain |
|
| esv3641561 | Chr.18:2395070 - 2566412 on Build GRCh38 | Loss |
|
| esv3893063 | Chr.18:2306414 - 2819763 on Build GRCh38 | Loss |
|
| nsv576310 | Chr.18:2278802 - 2566394 on Build GRCh38 | Gain |
|
| esv2761986 | Chr.18:2532931 - 2885250 on Build GRCh38 | Gain |
|
| nsv482935 | Chr.18:10001 - 2900002 on Build GRCh38 | Loss |
|
| nsv576251 | Chr.18:1700238 - 3043518 on Build GRCh38 | Gain |
|
| nsv576311 | Chr.18:2419932 - 2611877 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs66873847] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map