Assay Details
Target Gene Details
Entrez Gene ID: | 8792 |
Gene Name: | TNF receptor superfamily member 11a |
Gene Aliases: |
CD265, FEO, LOH18CR1, ODFR, OFE, OPTB7, OSTS, PDB2, RANK, TRANCER |
Location: |
Chr.18:62325287-62388096 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNFRSF11A | NM_001270949.1 | NP_001257878.1 | ||
| NM_001270950.1 | NP_001257879.1 | |||
| NM_001270951.1 | NP_001257880.1 | |||
| NM_001278268.1 | NP_001265197.1 | |||
| NM_003839.3 | NP_003830.1 | |||
| XM_011526244.2 | XP_011524546.1 | |||
| XM_011526245.2 | XP_011524547.1 | |||
| XM_017026064.1 | XP_016881553.1 | |||
| AB209762.1 | BAD92999.1 | |||
| AF018253.1 | AAB86809.1 | |||
| HE647782.1 | ||||
| HE649916.1 | ||||
| HE649917.1 | ||||
| HF584702.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs116333555] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map