Assay Details
Target Gene Details
Entrez Gene ID: | 8170 |
Gene Name: | solute carrier family 14 member 2 |
Gene Aliases: |
HUT2, UT-A2, UT2, UTA, UTR, hUT-A6 |
Location: |
Chr.18:45196357-45683686 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC14A2 | NM_001242692.1 | NP_001229621.1 | ||
| XM_017026015.1 | XP_016881504.1 | |||
| XM_017026016.1 | XP_016881505.1 | |||
| AK122698.1 | ||||
| AK292422.1 | ||||
| DB045789.1 |
Target Gene Details
Entrez Gene ID: | 101927980 |
Gene Name: | SLC14A2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.18:45438183-45507036 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC14A2-AS1 | NR_110899.1 | |||
| AK126075.1 | ||||
| BX109976.1 | ||||
| HY036726.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv275024 | Chr.18:45466850 - 45469907 on Build GRCh38 | Gain+Loss |
|
| nsv833638 | Chr.18:45304064 - 45494968 on Build GRCh38 | Gain |
|
| nsv525268 | Chr.18:45168400 - 45894830 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map