Assay Details
Target Gene Details
Entrez Gene ID: | 1829 |
Gene Name: | desmoglein 2 |
Gene Aliases: |
CDHF5, HDGC |
Location: |
Chr.18:31498004-31549008 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DSG2 | NM_001943.4 | NP_001934.2 | ||
| AK308408.1 | ||||
| BC099655.3 | AAH99655.1 | |||
| BC099656.3 | AAH99656.1 | |||
| BC099657.3 | AAH99657.1 | |||
| BP353513.1 | ||||
| CN347023.1 | ||||
| DC417739.1 | ||||
| HY268349.1 | ||||
| Z26317.1 | CAA81226.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833615 | Chr.18:31428433 - 31613013 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map