Assay Details
Target Gene Details
Entrez Gene ID: | 10982 |
Gene Name: | microtubule associated protein RP/EB family member 2 |
Gene Aliases: |
CSCSC2, EB1, EB2, RP1 |
Location: |
Chr.18:34976928-35143470 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAPRE2 | NM_001143826.2 | NP_001137298.1 | ||
| NM_001143827.2 | NP_001137299.1 | |||
| DC335558.1 | ||||
| DC341673.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs114031311] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map