Assay Details
Target Gene Details
Entrez Gene ID: | 28316 |
Gene Name: | cadherin 20 |
Gene Aliases: |
CDH7L3, Cdh7 |
Location: |
Chr.18:61333582-61555773 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDH20 | NM_031891.3 | NP_114097.2 | ||
| DA333316.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv577074 | Chr.18:61275224 - 61335721 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map