Assay Details
Target Gene Details
Entrez Gene ID: | 81035 |
Gene Name: | collectin subfamily member 12 |
Gene Aliases: |
CLP1, NSR2, SCARA4, SRCL |
Location: |
Chr.18:319355-500729 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COLEC12 | NM_130386.2 | NP_569057.1 | ||
| XM_011525741.2 | XP_011524043.1 | |||
| AB005145.1 | BAB72147.1 | |||
| AB034251.1 | BAD83592.1 | |||
| AB038518.1 | BAB39147.1 | |||
| AB052103.1 | BAB39148.1 | |||
| AL713657.1 | CAD28466.1 | |||
| BC060789.1 | AAH60789.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv576236 | Chr.18:319176 - 794272 on Build GRCh38 | Gain |
|
| nsv576235 | Chr.18:289153 - 407958 on Build GRCh38 | Gain |
|
| esv3641484 | Chr.18:340895 - 344933 on Build GRCh38 | Gain |
|
| dgv594e214 | Chr.18:340957 - 550862 on Build GRCh38 | Gain |
|
| nsv482935 | Chr.18:10001 - 2900002 on Build GRCh38 | Loss |
|
| nsv1066129 | Chr.18:258717 - 369148 on Build GRCh38 | Gain |
|
| nsv576228 | Chr.18:69836 - 1506323 on Build GRCh38 | Loss |
|
| esv3893056 | Chr.18:293519 - 463146 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73373535] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map