Assay Details
Target Gene Details
Entrez Gene ID: | 533 |
Gene Name: | ATPase H+ transporting V0 subunit b |
Gene Aliases: |
ATP6F, HATPL, VMA16 |
Location: |
Chr.1:43974648-43978300 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP6V0B | NM_001039457.2 | 1 | 79 | NP_001034546.1 |
| NM_001294333.1 | 1 | 79 | NP_001281262.1 | |
| NM_004047.4 | 1 | 79 | NP_004038.1 | |
| BP250477.1 | 1 | 79 |
More Information
Set Membership: |
|

Genomic Map