Assay Details
Target Gene Details
Entrez Gene ID: | 11332 |
Gene Name: | acyl-CoA thioesterase 7 |
Gene Aliases: |
ACH1, ACT, BACH, CTE-II, LACH, LACH1, hBACH |
Location: |
Chr.1:6264272-6393766 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACOT7 | NM_007274.3 | NP_009205.3 | ||
| NM_181864.2 | NP_863654.1 | |||
| NM_181865.2 | NP_863655.1 | |||
| NM_181866.2 | NP_863656.1 | |||
| AB074415.1 | BAC20174.1 | |||
| AB074416.1 | BAC20175.1 | |||
| AB074417.1 | BAC20176.1 | |||
| AB074418.1 | BAC20177.1 | |||
| AB074419.1 | BAC20178.1 | |||
| AK057168.1 | ||||
| AK289572.1 | ||||
| AK290097.1 | ||||
| AK291583.1 | ||||
| AK292202.1 | ||||
| AK300831.1 | ||||
| BC017365.2 | ||||
| BT006888.1 | AAP35534.1 | |||
| D88894.2 | BAA24350.1 | |||
| U91316.1 | AAB61211.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv965 | Chr.1:6253120 - 6297822 on Build GRCh38 | Deletion |
|
| nsv526371 | Chr.1:6247195 - 6307159 on Build GRCh38 | Loss |
|
| nsv545239 | Chr.1:6262876 - 6289815 on Build GRCh38 | Loss |
|
| nsv508825 | Chr.1:6221275 - 6286549 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map