Assay Details
Target Gene Details
Entrez Gene ID: | 79679 |
Gene Name: | V-set domain containing T cell activation inhibitor 1 |
Gene Aliases: |
B7-H4, B7H4, B7S1, B7X, B7h.5, PRO1291, VCTN1 |
Location: |
Chr.1:117143587-117210992 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VTCN1 | NM_001253849.1 | NP_001240778.1 | ||
| NM_001253850.1 | NP_001240779.1 | |||
| NM_024626.3 | NP_078902.2 | |||
| NR_045603.1 | ||||
| NR_045604.1 | ||||
| XM_011542143.2 | XP_011540445.2 | |||
| XM_017002335.1 | XP_016857824.1 | |||
| AK026071.1 | BAB15349.1 | |||
| AK225413.1 | ||||
| AK303466.1 | ||||
| AK310812.1 | ||||
| AY280972.1 | AAP37283.1 | |||
| AY346100.1 | AAQ24206.1 | |||
| AY358352.1 | AAQ88718.1 | |||
| BC065717.1 | AAH65717.1 | |||
| BC074729.2 | AAH74729.1 | |||
| BX648021.1 | ||||
| CN259811.1 | ||||
| DQ103757.1 | AAZ17406.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1009706 | Chr.1:117049372 - 117442154 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map