Assay Details
Target Gene Details
Entrez Gene ID: | 51100 |
Gene Name: | SH3 domain containing GRB2 like endophilin B1 |
Gene Aliases: |
Bif-1, CGI-61, PPP1R70, dJ612B15.2 |
Location: |
Chr.1:86704570-86748184 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SH3GLB1 | NM_001206651.1 | NP_001193580.1 | ||
| NM_001206652.1 | NP_001193581.1 | |||
| NM_001206653.1 | NP_001193582.1 | |||
| NM_016009.4 | NP_057093.1 | |||
| XM_006710672.2 | XP_006710735.1 | |||
| XM_006710673.2 | XP_006710736.1 | |||
| XM_011541538.1 | XP_011539840.1 | |||
| XM_017001408.1 | XP_016856897.1 | |||
| AB007960.2 | ||||
| AF151819.1 | AAD34056.1 | |||
| AF257318.1 | AAF81225.1 | |||
| AF263293.1 | AAF73017.1 | |||
| AF350371.1 | AAK27365.1 | |||
| AK001954.1 | BAA91999.1 | |||
| AK225864.1 | ||||
| AK303710.1 | ||||
| BC007455.2 | AAH07455.1 |
More Information
Set Membership: |
|

Genomic Map